Assay Details
| Gene Symbol: Stkld1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_198628.2 | NP_941030.2 | 7 - 8 | 569 | 78 | |
| GenBank mRNA | BC050806.1 | - | 7 - 8 | 570 | 78 | |
More Information
Updated Mapping Information
Set Membership: |
|

Stkld1
Genomic Map