Assay Details
| Gene Symbol: 0610009E02Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK075572.1 | - | 253 | 86 | ||
| BC023886.1 | - | 1 - 2 | 264 | 86 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | inositol polyphosphate-5-phosphatase E | Inpp5e |
1200002L24Rik, 72kDa, mKIAA0123 |
-36.7kb | Anti-sense | Chr. 2 - 26396249 - 26409219 on NCBI Build GRCm38 |
| Downstream | microRNA 6996 | Mir6996 |
Gm27531, mmu-mir-6996 |
12.1kb | Anti-sense | Chr. 2 - 26470059 - 26470119 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

0610009E02Rik
Genomic Map