Assay Details
| Gene Symbol: 0610009E02Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK075572.1 | - | 394 | 68 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | SEC16 homolog A (S. cerevisiae) | Sec16a |
AU024582, C230052J16Rik |
-11.5kb | Anti-sense | Chr. 2 - 26409431 - 26446162 on NCBI Build GRCm38 |
| Downstream | microRNA 6996 | Mir6996 |
Gm27531, mmu-mir-6996 |
11.3kb | Anti-sense | Chr. 2 - 26470059 - 26470119 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

0610009E02Rik
Genomic Map