Assay Details
| Gene Symbol: A230056P14Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_015495.2 | 4 - 5 | 520 | 148 | ||
| GenBank mRNA | AK043630.1 | - | 4 - 5 | 521 | 148 | |
| AK140288.1 | - | 4 - 5 | 521 | 148 | ||
| BC147226.1 | - | 2 - 3 | 159 | 148 | ||
| BC147227.1 | - | 2 - 3 | 159 | 148 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK140288.1 transcripts.
Set Membership: |
|

A230056P14Rik
Genomic Map