Assay Details
| Gene Symbol: Gm15927 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK051804.1 | - | 1 - 2 | 383 | 123 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
CCR4-NOT transcription complex, subunit 3 |
Cnot3 | A930039N10Rik | Anti-sense | Chr. 7 - 3644522 - 3661109 on NCBI Build GRCm38 | |
| Downstream | microRNA 3572 | Mir3572 |
mir-3572, mmu-mir-3572 |
11.3kb | Anti-sense | Chr. 7 - 3655962 - 3656047 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

Gm15927
Genomic Map