Assay Details
| Gene Symbol: Cybrd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028593.2 | NP_082869.2 | 1 - 2 | 259 | 59 | |
| GenBank mRNA | AF354666.1 | - | 1 - 2 | 201 | 59 | |
| AK008849.1 | - | 1 - 2 | 205 | 59 | ||
| AK029112.1 | - | 1 - 2 | 262 | 59 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cybrd1
Genomic Map