Assay Details
| Gene Symbol: Rsu1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009105.4 | NP_033131.2 | 1 - 2 | 219 | 58 | |
| GenBank mRNA | AK009288.1 | - | 1 - 2 | 112 | 58 | |
| AK011857.1 | - | 1 - 2 | 112 | 58 | ||
| AK135491.1 | - | 1 - 2 | 112 | 58 | ||
| AK137622.1 | - | 1 - 2 | 115 | 58 | ||
| AK149835.1 | - | 1 - 2 | 89 | 58 | ||
| AK150011.1 | - | 1 - 2 | 87 | 58 | ||
| AK152655.1 | - | 1 - 2 | 99 | 58 | ||
| AK169076.1 | - | 1 - 2 | 137 | 58 | ||
| AK172340.1 | - | 1 - 2 | 89 | 58 | ||
| BC003827.1 | - | 1 - 2 | 82 | 3493122 | 58 | |
| CX228720.1 | - | 1 - 2 | 219 | 58 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK011857.1, AK150011.1, AK152655.1, AK169076.1, BC003827.1, NM_009105.4 transcripts.
Set Membership: |
|

Rsu1
Genomic Map