Assay Details
| Gene Symbol: Capn6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007603.3 | NP_031629.3 | 6 - 7 | 1115 | 82 | |
| GenBank mRNA | AK145116.1 | - | 6 - 7 | 1130 | 82 | |
| BC052167.1 | - | 6 - 7 | 1016 | 6517565 | 82 | |
| Y12582.1 | - | 6 - 7 | 1024 | 82 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Capn6
Genomic Map