Assay Details
| Gene Symbol: Capn6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007603.3 | NP_031629.3 | 9 - 10 | 1498 | 86 | |
| GenBank mRNA | AK145116.1 | - | 9 - 10 | 1513 | 86 | |
| BC052167.1 | - | 9 - 10 | 1399 | 6517565 | 86 | |
| Y12582.1 | - | 9 - 10 | 1407 | 86 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Capn6
Genomic Map