Assay Details
| Gene Symbol: Il1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010554.4 | NP_034684.2 | 1 - 2 | 51 | 119 | |
| GenBank mRNA | AK156449.1 | - | 1 - 2 | 52 | 119 | |
| X01450.1 | - | 1 - 2 | 51 | 119 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK156449.1, NM_010554.4, X01450.1 transcripts.
Set Membership: |
|

Il1a
Genomic Map