Assay Details
| Gene Symbol: Il1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010554.4 | NP_034684.2 | 3 - 4 | 152 | 65 | |
| GenBank mRNA | AK156449.1 | - | 3 - 4 | 153 | 65 | |
| BC003727.1 | - | 2 - 3 | 121 | 3599550 | 65 | |
| X01450.1 | - | 3 - 4 | 152 | 65 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Il1a
Genomic Map