Assay Details
| Gene Symbol: Trip11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028446.1 | NP_082722.1 | 9 - 10 | 1657 | 136 | |
| GenBank mRNA | AK077954.1 | - | 9 - 10 | 1607 | 136 | |
| AK081763.1 | - | 9 - 10 | 1570 | 136 | ||
| AK148294.1 | - | 9 - 10 | 1633 | 136 | ||
| BC158098.1 | - | 9 - 10 | 1753 | 9088179 | 136 | |
More Information
Set Membership: |
|

Trip11
Genomic Map