Assay Details
| Gene Symbol: Atxn3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001167914.1 | NP_001161386.1 | 1 - 2 | 117 | 113 | |
| NM_029705.3 | NP_083981.2 | 1 - 2 | 117 | 113 | ||
| GenBank mRNA | AK008675.1 | - | 1 - 2 | 76 | 113 | |
| BC049743.1 | - | 1 - 2 | 117 | 113 | ||
| BC087880.1 | - | 1 - 2 | 112 | 6771498 | 113 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK008675.1, BC087880.1, NM_001167914.1, NM_029705.3 transcripts.
Set Membership: |
|

Atxn3
Genomic Map