Assay Details
| Gene Symbol: Atxn3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001167914.1 | NP_001161386.1 | 5 - 6 | 474 | 128 | |
| NM_029705.3 | NP_083981.2 | 5 - 6 | 474 | 128 | ||
| GenBank mRNA | AF537188.1 | - | 5 - 6 | 388 | 128 | |
| AK008675.1 | - | 5 - 6 | 433 | 128 | ||
| AK030842.1 | - | 5 - 6 | 414 | 128 | ||
| BC049743.1 | - | 5 - 6 | 474 | 128 | ||
| BC087880.1 | - | 5 - 6 | 469 | 6771498 | 128 | |
More Information
Updated Mapping Information
Set Membership: |
|

Atxn3
Genomic Map