Assay Details
| Gene Symbol: Cdc123 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133837.4 | NP_598598.1 | 10 - 11 | 765 | 140 | |
| GenBank mRNA | AK090122.1 | - | 10 - 11 | 766 | 140 | |
| AK141463.1 | - | 10 - 11 | 753 | 140 | ||
| AK159208.1 | - | 10 - 11 | 786 | 140 | ||
| BC019753.1 | - | 10 - 11 | 740 | 5008314 | 140 | |
| BC022972.1 | - | 10 - 11 | 754 | 5368775 | 140 | |
| BC024787.1 | - | 10 - 11 | 752 | 5359215 | 140 | |
More Information
Updated Mapping Information
Set Membership: |
|

Cdc123
Genomic Map