Assay Details
| Gene Symbol: Cdc123 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133837.4 | NP_598598.1 | 4 - 5 | 290 | 79 | |
| GenBank mRNA | AK090122.1 | - | 4 - 5 | 291 | 79 | |
| AK141463.1 | - | 4 - 5 | 278 | 79 | ||
| AK159208.1 | - | 4 - 5 | 311 | 79 | ||
| AK161246.1 | - | 4 - 5 | 277 | 79 | ||
| BC019753.1 | - | 4 - 5 | 265 | 5008314 | 79 | |
| BC022972.1 | - | 4 - 5 | 279 | 5368775 | 79 | |
| BC023821.1 | - | 4 - 5 | 282 | 5354711 | 79 | |
| BC024787.1 | - | 4 - 5 | 277 | 5359215 | 79 | |
More Information
Updated Mapping Information
Set Membership: |
|

Cdc123
Genomic Map