Assay Details
| Gene Symbol: Upf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081132.1 | NP_001074601.1 | 10 - 11 | 2168 | 121 | |
| GenBank mRNA | BC138358.1 | - | 10 - 11 | 2131 | 8861378 | 121 |
| BC138359.1 | - | 10 - 11 | 2131 | 8861379 | 121 | |
More Information
Set Membership: |
|

Upf2
Genomic Map