Assay Details
| Gene Symbol: Dhtkd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081131.2 | NP_001074600.1 | 7 - 8 | 1483 | 65 | |
| GenBank mRNA | AK049272.1 | - | 1474 | 65 | ||
| AK173223.1 | - | 2 - 3 | 541 | 65 | ||
| BC117994.1 | - | 1434 | 40097991 | 65 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Dhtkd1
Genomic Map