Assay Details
| Gene Symbol: Anapc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175300.4 | NP_780509.2 | 1 - 2 | 210 | 108 | |
| GenBank mRNA | AK033784.1 | - | 1 - 2 | 178 | 108 | |
| AK129351.1 | - | 1 - 2 | 163 | 108 | ||
| AK154465.1 | - | 1 - 2 | 164 | 108 | ||
| AK170676.1 | - | 1 - 2 | 176 | 108 | ||
| BC027351.1 | - | 1 - 2 | 158 | 108 | ||
| BC137583.1 | - | 1 - 2 | 178 | 8860602 | 108 | |
| CJ164053.1 | - | 1 - 2 | 210 | 108 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Anapc2
Genomic Map