Assay Details
| Gene Symbol: Lpcat4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_207206.2 | NP_997089.1 | 12 - 13 | 1345 | 62 | |
| GenBank mRNA | BC046276.1 | - | 10 - 11 | 1232 | 62 | |
| BC068131.1 | - | 12 - 13 | 1345 | 6414152 | 62 | |
| BC080829.1 | - | 12 - 13 | 1312 | 30435722 | 62 | |
More Information
Updated Mapping Information
Set Membership: |
|

Lpcat4
Genomic Map