Assay Details
| Gene Symbol: Adtrp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001145875.1 | NP_001139347.1 | 1 - 2 | 122 | 64 | |
| GenBank mRNA | AK079178.1 | - | 1 - 2 | 123 | 64 | |
| AK135327.1 | - | 1 - 2 | 123 | 64 | ||
| BY719442.1 | - | 1 - 2 | 119 | 64 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK135327.1, NM_001145875.1 transcripts.
Set Membership: |
|

Adtrp
Genomic Map