Assay Details
| Gene Symbol: Rcor1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_198023.2 | NP_932140.1 | 7 - 8 | 650 | 87 | |
| GenBank mRNA | AB093210.1 | - | 3 - 4 | 351 | 87 | |
| AK077445.1 | - | 1 - 2 | 91 | 87 | ||
| BC042731.1 | - | 7 - 8 | 650 | 4936863 | 87 | |
More Information
Updated Mapping Information
Set Membership: |
|

Rcor1
Genomic Map