Assay Details
| Gene Symbol: Rcor1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_198023.2 | NP_932140.1 | 10 - 11 | 976 | 65 | |
| GenBank mRNA | AB093210.1 | - | 6 - 7 | 677 | 65 | |
| AK077445.1 | - | 4 - 5 | 417 | 65 | ||
| BC042731.1 | - | 10 - 11 | 976 | 4936863 | 65 | |
More Information
Updated Mapping Information
Set Membership: |
|

Rcor1
Genomic Map