Assay Details
| Gene Symbol: Traf3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001286122.1 | NP_001273051.1 | 5 - 6 | 817 | 64 | |
| NM_011632.3 | NP_035762.2 | 6 - 7 | 928 | 64 | ||
| GenBank mRNA | AK147304.1 | - | 5 - 6 | 745 | 64 | |
| AK147362.1 | - | 5 - 6 | 818 | 64 | ||
| BC137634.1 | - | 5 - 6 | 643 | 8860653 | 64 | |
| BC137635.1 | - | 5 - 6 | 643 | 8860654 | 64 | |
| U21050.1 | - | 6 - 7 | 747 | 64 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Traf3
Genomic Map