Assay Details
| Gene Symbol: Traf3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011632.3 | NP_035762.2 | 7 - 8 | 1011 | 93 | |
| GenBank mRNA | AK148231.1 | - | 1 - 2 | 167 | 93 | |
| BC137634.1 | - | 6 - 7 | 726 | 8860653 | 93 | |
| BC137635.1 | - | 6 - 7 | 726 | 8860654 | 93 | |
| U21050.1 | - | 7 - 8 | 830 | 93 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Traf3
Genomic Map