Assay Details
| Gene Symbol: Traf3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011632.3 | NP_035762.2 | 8 - 9 | 1086 | 69 | |
| GenBank mRNA | AK148231.1 | - | 2 - 3 | 242 | 69 | |
| BC137634.1 | - | 7 - 8 | 801 | 8860653 | 69 | |
| BC137635.1 | - | 7 - 8 | 801 | 8860654 | 69 | |
| U21050.1 | - | 8 - 9 | 905 | 69 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Traf3
Genomic Map