Assay Details
| Gene Symbol: Ralgds | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001145834.1 | NP_001139306.1 | 15 - 16 | 2366 | 84 | |
| NM_001145835.1 | NP_001139307.1 | 15 - 16 | 2278 | 84 | ||
| NM_001145836.1 | NP_001139308.1 | 15 - 16 | 2242 | 84 | ||
| NM_009058.2 | NP_033084.2 | 15 - 16 | 2402 | 84 | ||
| GenBank mRNA | AK129328.1 | - | 15 - 16 | 2243 | 84 | |
| AK141959.1 | - | 15 - 16 | 2233 | 84 | ||
| AK149644.1 | - | 10 - 11 | 1137 | 84 | ||
| AK159020.1 | - | 15 - 16 | 2235 | 84 | ||
| AK159754.1 | - | 15 - 16 | 2248 | 84 | ||
| BC027225.1 | - | 15 - 16 | 2060 | 3486732 | 84 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ralgds
Genomic Map