Assay Details
| Gene Symbol: Ralgds | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001145834.1 | NP_001139306.1 | 8 - 9 | 1676 | 65 | |
| NM_001145835.1 | NP_001139307.1 | 8 - 9 | 1588 | 65 | ||
| NM_001145836.1 | NP_001139308.1 | 8 - 9 | 1552 | 65 | ||
| NM_009058.2 | NP_033084.2 | 8 - 9 | 1712 | 65 | ||
| GenBank mRNA | AK129328.1 | - | 8 - 9 | 1553 | 65 | |
| AK141959.1 | - | 8 - 9 | 1543 | 65 | ||
| AK149644.1 | - | 3 - 4 | 447 | 65 | ||
| AK159020.1 | - | 8 - 9 | 1545 | 65 | ||
| AK159754.1 | - | 8 - 9 | 1558 | 65 | ||
| BC027225.1 | - | 8 - 9 | 1370 | 3486732 | 65 | |
| L07924.1 | - | 8 - 9 | 1548 | 65 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ralgds
Genomic Map