Assay Details
| Gene Symbol: Plxdc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026162.6 | NP_080438.2 | 12 - 13 | 2135 | 67 | |
| GenBank mRNA | AF378761.1 | - | 12 - 13 | 1334 | 67 | |
| AK004640.1 | - | 12 - 13 | 2117 | 67 | ||
| AK019482.2 | - | 12 - 13 | 2093 | 67 | ||
| AK035165.1 | - | 12 - 13 | 2137 | 67 | ||
| AK154921.1 | - | 12 - 13 | 2082 | 67 | ||
| BC057881.1 | - | 12 - 13 | 1395 | 5349869 | 67 | |
More Information
Updated Mapping Information
Set Membership: |
|

Plxdc2
Genomic Map