Assay Details
| Gene Symbol: Plxdc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026162.6 | NP_080438.2 | 1 - 2 | 934 | 66 | |
| GenBank mRNA | AF378761.1 | - | 1 - 2 | 133 | 66 | |
| AK004640.1 | - | 1 - 2 | 916 | 66 | ||
| AK019482.2 | - | 1 - 2 | 890 | 66 | ||
| AK035165.1 | - | 1 - 2 | 935 | 66 | ||
| AK154921.1 | - | 1 - 2 | 881 | 66 | ||
| BC057881.1 | - | 1 - 2 | 194 | 5349869 | 66 | |
More Information
Updated Mapping Information
Set Membership: |
|

Plxdc2
Genomic Map