Assay Details
| Gene Symbol: Ndufaf5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027093.4 | NP_081369.2 | 4 - 5 | 443 | 132 | |
| GenBank mRNA | BC005630.1 | - | 4 - 5 | 310 | 132 | |
| CD363717.1 | - | 4 - 5 | 413 | 132 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the BC005630.1 transcripts.
Set Membership: |
|

Ndufaf5
Genomic Map