Assay Details
| Gene Symbol: Spata7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001289572.1 | NP_001276501.1 | 5 - 6 | 882 | 82 | |
| NM_001289574.1 | NP_001276503.1 | 6 - 7 | 1044 | 82 | ||
| NM_178914.4 | NP_849245.2 | 6 - 7 | 978 | 82 | ||
| GenBank mRNA | AK036359.1 | - | 6 - 7 | 962 | 82 | |
| AK077500.1 | - | 6 - 7 | 889 | 82 | ||
| AK161308.1 | - | 5 - 6 | 828 | 82 | ||
| BC046960.1 | - | 6 - 7 | 936 | 6391450 | 82 | |
| DQ196101.1 | - | 5 - 6 | 708 | 82 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Spata7
Genomic Map