Assay Details
| Gene Symbol: Spata7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001289572.1 | NP_001276501.1 | 6 - 7 | 955 | 75 | |
| NM_001289574.1 | NP_001276503.1 | 7 - 8 | 1117 | 75 | ||
| NM_178914.4 | NP_849245.2 | 7 - 8 | 1051 | 75 | ||
| GenBank mRNA | AK036359.1 | - | 7 - 8 | 1035 | 75 | |
| AK077500.1 | - | 7 - 8 | 962 | 75 | ||
| AK161308.1 | - | 6 - 7 | 901 | 75 | ||
| BC046960.1 | - | 7 - 8 | 1009 | 6391450 | 75 | |
| DQ196101.1 | - | 6 - 7 | 781 | 75 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Spata7
Genomic Map