Assay Details
| Gene Symbol: Naa38 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_030083.2 | NP_084359.1 | 1 - 2 | 185 | 64 | |
| GenBank mRNA | AK018782.1 | - | 1 - 2 | 186 | 64 | |
| BC038659.1 | - | 1 - 2 | 139 | 64 | ||
| BC062267.1 | - | 1 - 2 | 155 | 1532489 | 64 | |
More Information
Updated Mapping Information
Set Membership: |
|

Naa38
Genomic Map