Assay Details
| Gene Symbol: Kcnab3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010599.4 | NP_034729.3 | 2 - 3 | 709 | 95 | |
| GenBank mRNA | AK031806.1 | - | 2 - 3 | 690 | 95 | |
| AK044102.1 | - | 2 - 3 | 664 | 95 | ||
| AK046194.1 | - | 2 - 3 | 699 | 95 | ||
| AK049316.1 | - | 2 - 3 | 710 | 95 | ||
| AK083134.1 | - | 2 - 3 | 711 | 95 | ||
| BC017518.1 | - | 2 - 3 | 635 | 4507029 | 95 | |
More Information
Updated Mapping Information
Set Membership: |
|

Kcnab3
Genomic Map