Assay Details
| Gene Symbol: Kcnab3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010599.4 | NP_034729.3 | 4 - 5 | 821 | 79 | |
| GenBank mRNA | AK029750.1 | - | 3 - 4 | 410 | 79 | |
| AK031806.1 | - | 4 - 5 | 802 | 79 | ||
| AK044102.1 | - | 4 - 5 | 776 | 79 | ||
| AK046194.1 | - | 4 - 5 | 811 | 79 | ||
| AK049316.1 | - | 4 - 5 | 822 | 79 | ||
| AK083134.1 | - | 4 - 5 | 823 | 79 | ||
| BC017518.1 | - | 4 - 5 | 747 | 4507029 | 79 | |
More Information
Updated Mapping Information
Set Membership: |
|

Kcnab3
Genomic Map