Assay Details
| Gene Symbol: Kcnab3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010599.4 | NP_034729.3 | 6 - 7 | 916 | 77 | |
| GenBank mRNA | AK029750.1 | - | 5 - 6 | 505 | 77 | |
| AK031806.1 | - | 6 - 7 | 897 | 77 | ||
| AK044102.1 | - | 6 - 7 | 871 | 77 | ||
| AK046194.1 | - | 6 - 7 | 906 | 77 | ||
| AK049316.1 | - | 6 - 7 | 917 | 77 | ||
| AK083134.1 | - | 6 - 7 | 918 | 77 | ||
| BC017518.1 | - | 6 - 7 | 842 | 4507029 | 77 | |
More Information
Updated Mapping Information
Set Membership: |
|

Kcnab3
Genomic Map