Assay Details
| Gene Symbol: Atp1b2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013415.5 | NP_038201.1 | 4 - 5 | 1146 | 62 | |
| GenBank mRNA | AK141786.1 | - | 4 - 5 | 1147 | 62 | |
| AK158321.1 | - | 4 - 5 | 1149 | 62 | ||
| BC034586.1 | - | 4 - 5 | 1128 | 62 | ||
| BC042467.1 | - | 4 - 5 | 1015 | 4500972 | 62 | |
| BC058763.1 | - | 4 - 5 | 1104 | 6490870 | 62 | |
| X16645.1 | - | 4 - 5 | 702 | 62 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Atp1b2
Genomic Map