Assay Details
| Gene Symbol: Atp1b2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013415.5 | NP_038201.1 | 5 - 6 | 1193 | 64 | |
| GenBank mRNA | AK141786.1 | - | 5 - 6 | 1194 | 64 | |
| AK158321.1 | - | 5 - 6 | 1196 | 64 | ||
| BC042467.1 | - | 5 - 6 | 1062 | 4500972 | 64 | |
| BC058763.1 | - | 5 - 6 | 1151 | 6490870 | 64 | |
| X16645.1 | - | 5 - 6 | 749 | 64 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Atp1b2
Genomic Map