Assay Details
| Gene Symbol: Cdc42ep5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_021454.3 | NP_067429.1 | 1 - 2 | 245 | 85 | |
| GenBank mRNA | AF164119.1 | - | 1 - 2 | 45 | 85 | |
| AK008154.1 | - | 1 - 2 | 245 | 85 | ||
| BC006758.1 | - | 1 - 2 | 71 | 3710012 | 85 | |
| BC103774.1 | - | 1 - 2 | 256 | 6529557 | 85 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AF164119.1, AK008154.1, BC006758.1, BC103774.1, NM_021454.3 transcripts.
Set Membership: |
|

Cdc42ep5
Genomic Map