Assay Details
| Gene Symbol: Rdh13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001290409.1 | NP_001277338.1 | 1 - 2 | 215 | 77 | |
| NM_175372.4 | NP_780581.1 | 1 - 2 | 215 | 77 | ||
| GenBank mRNA | AK028434.1 | - | 1 - 2 | 208 | 77 | |
| AK034180.1 | - | 160 | 77 | |||
| BC082583.1 | - | 1 - 2 | 167 | 30613493 | 77 | |
| BY083213.1 | - | 1 - 2 | 216 | 77 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Rdh13
Genomic Map