Assay Details
| Gene Symbol: Rdh13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001290409.1 | NP_001277338.1 | 2 - 3 | 331 | 69 | |
| NM_001290411.1 | NP_001277340.1 | 2 - 3 | 364 | 69 | ||
| NM_175372.4 | NP_780581.1 | 2 - 3 | 331 | 69 | ||
| GenBank mRNA | AK028434.1 | - | 2 - 3 | 324 | 69 | |
| AK034180.1 | - | 276 | 69 | |||
| AK089458.1 | - | 2 - 3 | 737 | 69 | ||
| BC082583.1 | - | 2 - 3 | 283 | 30613493 | 69 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001290411.1 transcripts.
Set Membership: |
|

Rdh13
Genomic Map