Assay Details
| Gene Symbol: Rdh13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001290411.1 | NP_001277340.1 | 3 - 4 | 520 | 63 | |
| NM_175372.4 | NP_780581.1 | 3 - 4 | 487 | 63 | ||
| GenBank mRNA | AK028434.1 | - | 3 - 4 | 480 | 63 | |
| BC082583.1 | - | 3 - 4 | 439 | 30613493 | 63 | |
More Information
Set Membership: |
|

Rdh13
Genomic Map