Assay Details
| Gene Symbol: Ncr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010746.3 | NP_034876.2 | 2 - 3 | 95 | 92 | |
| GenBank mRNA | AJ223765.1 | - | 2 - 3 | 84 | 92 | |
| AK079401.1 | - | 2 - 3 | 96 | 92 | ||
| BC042788.1 | - | 2 - 3 | 65 | 92 | ||
| BC115364.2 | - | 2 - 3 | 96 | 40045898 | 92 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ncr1
Genomic Map