Assay Details
| Gene Symbol: Ncr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AJ223765.1 | - | 6 - 7 | 748 | 128 | |
| AK172251.1 | - | 5 - 6 | 792 | 128 | ||
| BC042788.1 | - | 6 - 7 | 729 | 128 | ||
| BC115364.2 | - | 6 - 7 | 760 | 40045898 | 128 | |
More Information
Set Membership: |
|

Ncr1
Genomic Map