Assay Details
| Gene Symbol: Hnmt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_080462.2 | NP_536710.1 | 2 - 3 | 211 | 77 | |
| GenBank mRNA | AB070524.1 | - | 2 - 3 | 207 | 77 | |
| AB444950.1 | - | 1 - 2 | 141 | 77 | ||
| AB444951.1 | - | 1 - 2 | 141 | 77 | ||
| AF297037.1 | - | 2 - 3 | 211 | 77 | ||
| AK050129.1 | - | 2 - 3 | 234 | 77 | ||
| AK160131.1 | - | 1 - 2 | 155 | 77 | ||
| BC033928.1 | - | 2 - 3 | 203 | 1546890 | 77 | |
More Information
Updated Mapping Information
Set Membership: |
|

Hnmt
Genomic Map