Assay Details
| Gene Symbol: Psd4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177611.3 | NP_808279.1 | 3 - 4 | 1214 | 61 | |
| GenBank mRNA | AK043669.1 | - | 4 - 5 | 1506 | 61 | |
| AK150238.1 | - | 3 - 4 | 1243 | 61 | ||
| AK169859.1 | - | 3 - 4 | 1243 | 61 | ||
| BC068231.1 | - | 3 - 4 | 1214 | 30361859 | 61 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK150238.1 transcripts.
Set Membership: |
|

Psd4
Genomic Map