Assay Details
| Gene Symbol: Pak6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033254.3 | NP_001028426.2 | 3 - 4 | 640 | 102 | |
| NM_001145854.1 | NP_001139326.1 | 2 - 3 | 165 | 102 | ||
| GenBank mRNA | AK028788.1 | - | 2 - 3 | 149 | 102 | |
| AK145601.1 | - | 2 - 3 | 139 | 102 | ||
| AK158913.1 | - | 3 - 4 | 914 | 102 | ||
| AK158944.1 | - | 3 - 4 | 914 | 102 | ||
| BC059912.1 | - | 3 - 4 | 447 | 102 | ||
| BC079572.1 | - | 3 - 4 | 640 | 102 | ||
| BC150754.1 | - | 3 - 4 | 530 | 9087665 | 102 | |
| BY035804.1 | - | 2 - 3 | 166 | 102 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK145601.1, AK158913.1, AK158944.1, BC150754.1, NM_001033254.3, NM_001145854.1 transcripts.
Set Membership: |
|

Pak6
Genomic Map