Assay Details
| Gene Symbol: Bub1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009773.3 | NP_033903.2 | 12 - 13 | 1781 | 118 | |
| GenBank mRNA | AF107296.1 | - | 12 - 13 | 1736 | 118 | |
| BC031577.1 | - | 12 - 13 | 1732 | 4953223 | 118 | |
| BU054510.1 | - | 3 - 4 | 283 | 118 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Bub1b
Genomic Map