Assay Details
| Gene Symbol: Bub1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009773.3 | NP_033903.2 | 16 - 17 | 2341 | 64 | |
| GenBank mRNA | AK133909.1 | - | 16 - 17 | 2282 | 64 | |
More Information
Set Membership: |
|

Bub1b
Genomic Map